missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin E2/PN1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
£146.86 - £464.80
Specifications
Antigen | Serpin E2/PN1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18706367
|
R&D Systems
AF2980-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18049971
|
R&D Systems
AF2980 |
100 μg |
£464.80
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin E2/PN1 Polyclonal specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blot.Specifications
Serpin E2/PN1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2 | |
SERPINE2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001130000 | |
5270 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin E2 Met1-Pro397 Accession # NP_001130000 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Serpin E2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Serpin E2 is observed and less than 1% cross-reactivity with recombinant human (rh) Serpin E1, rhSerpin I1, rhSerpin I2, and rhSerpin B6 is observed. |
For Research Use Only