missing translation for 'onlineSavingsMsg'
Learn More
Sesamol, 98%, Thermo Scientific Chemicals Product Code.: 10193370

Sesamol, 98%, Thermo Scientific Chemicals

Product Code. 10193370
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10193370

Brand: Thermo Scientific Acros 132690250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00005827
InChI Key LUSZGTFNYDARNI-UHFFFAOYSA-N
Synonym sesamol, 3,4-methylenedioxy phenol, 5-hydroxy-1,3-benzodioxole, 3,4-methylenedioxyphenol, 2h-1,3-benzodioxol-5-ol, 3,4-methylendioxyphenol, methylene ether of oxyhydroquinone, phenol, 3,4-methylenedioxy, 5-benzodioxolol, unii-94iea0nv89
PubChem CID 68289
ChEBI CHEBI:9126
IUPAC Name 1,3-benzodioxol-5-ol
SMILES C1OC2=C(O1)C=C(C=C2)O

Specifications

Melting Point 63.0°C to 65.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 19, 66
Solubility Information Solubility in water: slightly soluble.
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sesamol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0408342 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number132690050,132690250,132691000
Lot NumberA0446328 Certificate TypeCertificate of Analysis Date17/10/2022 Catalog Number132690050,132690250,132691000
Lot NumberA0414790 Certificate TypeCertificate of Analysis Date16/06/2022 Catalog Number132690050,132690250,132691000
Lot NumberA0434082 Certificate TypeCertificate of Analysis Date14/09/2021 Catalog Number132690050,132690250,132691000
Product Identifier
  • Sesamol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sesamol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.