Learn More
Grant Instruments™ Liquid McFarland Standards Set
For use with DEN-1 and DEN-1B Densitometers. Grant Instruments™ Liquid McFarland Standards Set is supplied in 16mm dia. glass tubes.
Brand:  Grant Instruments™ DEN MCF STDS
436.38 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 13160623
  • £503.00 / 5 sets
Explore more special offers
Description
Description
  • Supplied in 16mm ø glass tubes.
  • 0.5 / 1.0 / 2.0 / 3.0 / 4.0 can be decanted into an alternative tube (shelf-life no longer valid).
  • Requires D16 adaptor, supplied with DEN-1 / DEN-1B
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEN-1 & DEN-1B Densitometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Liquid McFarland Standards Set > For Use With: DEN-1 & DEN-1B Densitometer