Learn More
MP Biomedicals™ Set of 4 dNTPs (100mm)
Offers maximum flexibility in preparation of reaction mixes. MP Biomedicals Set of 4 dNTPs (100mm) includes four separate vials of highly purified sodium salts of dATP, dCTP, dGTP and dTTP. These solutions achieve sensitive and consistent PCR results.
Brand:  MP Biomedicals™ 11NTACG111
 View more versions of this product
Product Code. 11467110
  • £515.00 / Pack of 4
Explore more special offers
Description
Description
This product allows the addition of each dNTP to be altered as required for specific experiments, such as:
  • When adding a radioactive nucleotide for labeling by PCR
  • To increase the chance of non-specific mutations
  • In PCR, real-time PCR, cDNA synthesis and DNA sequencing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Set 4 DNTPs composed of four separate vials of sodium salts of dATP, dCTP, dGTP and dTTP. Store at -20°C.
4 x 100 μmol
100 mM
99+%
Standard PCR, Real Time PCR (qPCR), RT-PCR