Learn More
Heidolph Set Glass Sensor
Modular temperature sensor consisting of a standard PT1000 (V4A) and a glass coating when working with aggressive media.
Brand: Heidolph
Additional Details : Weight : 0.20000kg
Description
- Modular temperature sensor consisting of a standard PT1000 (V4A) and a glass coating when working with aggressive media.
- Compared to other glass sensors, you will only have to change the glass coating when it comes to glass breakage and not the whole temperature sensor.
- Compatible with: Hei-PLATE Mix 'n' Heat Core+, Expert, Ultimate
Specifications
Temperature Probe | |
2 Years | |
Plastic |