Learn More
BRAND™ Set of Special Pegs
Set of Special Pegs for use with a draining rack.
Brand:  BRAND™ 95111
Product Code. 10401521
  • £23.50 / Pack of 11
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For narrow-mouthed or small vessels
  • Can be used with test tubes, small volumetric flasks, and graduated cylinders
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
11/Pk
6 mm
Drain Rack
100 mm