Learn More
Interscience™ Set of Tygon™ Tubings for Pump Head
20 cm long Tygon™ tubings for use with DiluFlow and FlexiPump pump heads.
Brand: Interscience™ 561304
This item is not returnable.
View return policy
Description
- Transparent
- Flexible Tygon™ tubing
Specifications
Pump Tubing | |
Adding Tubing to Dilutor Pumps | |
6 Tubings/Set |
Autoclavable | |
DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Interscience™ Set of Tygon™ Tubings for Pump Head > Set of Tygon Tubings for Pump Head; For Use With (Equipment): DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro
Spot an opportunity for improvement?Share a Content Correction