missing translation for 'onlineSavingsMsg'
Learn More

Seward™ Stomacher™ Bag Racks

Product Code. p-149238
Click to view available options
Holds:
Bags
For Use With (Equipment):
Stomacher 3500
Stomacher 400
Stomacher 400 lab blenders
Stomacher 80Biomaster
Dimensions (L x W x H):
15 cm x 42 cm x 35 cm
30 x 11 x 12 cm
35 cm x 10 cm x 22 cm
8 cm x 10.5 cm x 19 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10319227

Brand: Seward™ BA6091

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Seward has developed a full range of Stomacher™ Accessories to help provide a safe and efficient working environment in your laboratory.

Help to organize sample bags

  • High-grade plastic polymer construction
  • Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
  • Perfect for loading closure bags pre-enrichment into the incubator
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)

Specifications

Product Type Bag Rack
Autoclavable Not Autoclavable
Color Clear
Holds Bags
Material Polymer
Positions 1
Sterility Non-sterile
For Use With (Application) To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment) Stomacher 400 lab blenders
No. per Pack 1
Quantity 1
Dimensions (L x W x H) 8 cm x 10.5 cm x 19 cm
Weight (Metric) 0.4kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.