missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Sf-900™ II SFM (1X)
Sf-900™ II SFM (1X)
Brand: Gibco™ 10902153
Code : 50
Additional Details : Weight : 0.50000kg
Description
Sf-900™ II SFM is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells and for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.Gibco™ Sf-900™ II SFM features:
Superior Long-Term, High Density Growth
Spodoptera frugiperda (Sf9) cells grown in Sf-900™ II SFM achieve maximum cell densities of 9 to 12 × 106 cells/mL, a significant improvement over competitor's formulations and Grace’s medium (see Product Manual). Increases in maximum cell densities of 20-100% are also observed with the Lymantria dispar (Gypsy moth) and Trichoplusia ni (Tn-368; Cabbage Looper) cell lines. Gibco™ Sf-900™ II SFM is capable of supporting the cultures to >20 passages.
Optimized for Recombinant Protein Production
Traditionally, Grace's medium supplemented with 10% FBS has been used for recombinant protein expression. Sf-900™ II SFM is an improved serum-free, protein-free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect virus and rDNA proteins.
Serum-Free, Protein-Free, Ready-to-Use Formulation
Gibco™ Sf-900™ II SFM is a serum-free, protein-free medium that allows for much easier purification of your protein of interest. Sf-900™ II SFM is ready to use; it does not require addition of serum, glutamine or surfactants. Cells adapted to other commercially available serum-free media can be subcultured directly into Sf-900™ II SFM, usually without any further adaptation. Cells usually require some adaptation from serum-containing formulations.
Scalable in Bioreactors
Utility of Sf-900™ II SFM in larger scale cell culture systems was demonstrated with a 5L Celligen™ bioreactor. Successful infections with rAcNPV were carried out producing rβ-Gal and rEPO (see Product Manual).
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use. Customers using Gibco™ Sf-900 II SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).
cGMP Manufacturing and Quality System
Gibco™ Sf-900 II SFM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Specifications
ISO 13485 standards | |
Sf-900™ II SFM (1 X), 10 x 500mL, Liquid, Cell Culture, Spodoptera Frugiperda, S. Frugiperda Species, Sf21, Sf9 Cell Line, Insect Cells Cell Type, L-Glutamine, Serum-Free, Serum-Free, Protein-Free Classification, For Insect Cell Culture | |
Liquid | |
Cell culture, insect cell culture, bioproduction, viral vaccine production, Sf9 and Sf21 cell culture |
2 to 8°C | |
Insect Cell | |
Insect Cell Serum Free Medium (SFM) | |
10 x 500 mL |
For Research Use Only. Not for use in diagnostic procedures.