missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SFM4Transfx-293 Media
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
£31.60 - £317.90
Specifications
Form | Liquid |
---|---|
Product Type | SFM4Transfx-293 |
Certifications/Compliance | cGMP |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10257653
|
Cytiva
SH30860.LS |
6 x 1000 mL | N/A | N/A | N/A | |||||
10649783
|
Cytiva
SH30860.01 |
500 mL | N/A | N/A | N/A | |||||
11536341
|
Cytiva
SH30860.02 |
1000 mL |
£66.71
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- ADCF
- Supports high transfection efficiency
- Metabolically designed for high cell yield and recombinant protein/vector production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
Liquid | |
SFM4Transfx-293 | |
cGMP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva SFM4Transfx-293 Media