missing translation for 'onlineSavingsMsg'
Learn More

SFRS9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17926-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877934

  • £130.00 / 20µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the serine/arginine (SR)-rich family of pre-mRNA splicing factors, which constitute part of the spliceosome. Each of these factors contains an RNA recognition motif (RRM) for binding RNA and an RS domain for binding other proteins. The RS domain is rich in serine and arginine residues and facilitates interaction between different SR splicing factors. In addition to being critical for mRNA splicing, the SR proteins have also been shown to be involved in mRNA export from the nucleus and in translation. Two pseudogenes, one on chromosome 15 and the other on chromosome 21, have been found for this gene.
Specifications

Specifications

SFRS9
Polyclonal
Unconjugated
SRSF9
SFRS9, SRp30c, SRSF9
Rabbit
Antigen Affinity Chromatography
RUO
108014, 288701, 8683
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13242, Q5PPI1, Q9D0B0
Srsf9
SFRS9 Fusion Protein Ag12347
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers