Learn More
Tocris Bioscience™ SGC GAK 1
High affinity cyclin G associated kinase (GAK) inhibitor
Brand:  Tocris Bioscience™ 6564/25
 View more versions of this product
Product Code. 18715348
  • £314.25 / 25mg
Explore more special offers
Description
Description
SGC GAK 1 is a high affinity cyclin G associated kinase (GAK) inhibitor (Kd = 1.9 nM). Exhibits >30-fold selectivity for GAK over a panel of other kinases. Inhibits GAK in live cell assays (IC50 = 110 nM).
Specifications
Specifications
6-Bromo-N-(3,4,5-trimethoxyphenyl)-4-quinolinamine
25 mg
99%
2226517-76-4
C18H17BrN2O3