missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ SGC Inlet Split/Splitless Focus Liner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 092058

Product Code. 12913991

  • £194.00 / Pack of 5
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimadzu instruments
Specifications

Specifications

5/Pk.
Wool
3.4 mm
95 mm
Inlet Split/Splitless Focus Liner
5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.