Learn More
Trajan Scientific and Medical™ SGE Analytical Science Quartz Wool
GC inlet liner for use with Pyrojector II and pyrolysis chromatography.
Brand: Trajan Scientific and Medical™ 18060001
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- High Resolution Capillary GC
- For use with Pyrolysis chromatography applications
- Works in conjunction with Pyrojector II which thermally fragments essentially non-volatile molecules into volatile fragments, making them suitable for chromatographic analysis
Specifications
GC Inlet Liner | |
Quartz | |
Quartz Wool | |
0.5 g |