Learn More
Trajan™ CTC PAL Removable Needle Syringe
Equipped with PTFE-tipped plunger. For use with HPLC Autosamplers
Brand:  Trajan™ 006330
Product Code. 12907566
  • £90.00 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for High performance liquid chromatology (HPLC)
Specifications
Specifications
PTFE (Plunger)
0.37 mm
HPLC Autosampler Syringe
LC
22 G
0.72 mm
250 μL