Learn More
Trajan™ Diamond MS Autosampler Syringes: For Agilent
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™ 002760
 View more versions of this product
Product Code. 13147976
  • £91.10 / Each
Explore more special offers
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Improves mass spectrometry analysis sensitivity
10R-AG-GT-MS1
10 μL
Diamond MS Syringe
Ea.
Removable