Learn More
Trajan™ Diamond MS Manual Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand: Trajan™ 005750
This item is not returnable.
View return policy
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Removable ultra smooth needle | |
100R-GT-MS1 | |
100 μL |
Diamond MS Manual Syringe | |
Ea. | |
Removable |