Learn More
Trajan™ Diamond MS Manual Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand:  Trajan™ 006750
 View more versions of this product
Product Code. 13127976
  • £84.60 / Each
Explore more special offers
Description
Description
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Specifications
Removable ultra smooth needle
250R-GT-MS1
250 μL
Diamond MS Manual Syringe
Ea.
Removable