Learn More
Trajan™ eVol™ XCHANGE Syringes
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
Brand: Trajan™ 2910329
This item is not returnable.
View return policy
Beschreibung
- Liquid handling user independent device
- Reduction in need for re-analysis
- Syringes easily and quickly changed
- Replacement parts available
Spezifikation
25 G | |
1.9 in. | |
eVol™ Syringe | |
100 μL | |
Bevel |
Liquid handling procedure | |
50 mm | |
3/Pk. | |
50 mm |