Learn More
Trajan™ Hexnut Fittings
HexNut fittings provide ideal connections in HPLC systems
£35.20 - £153.00
Specifications
Thread Size 10-32
Material Stainless Steel
Pressure 5000 psi
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
15249103
View Documents
Trajan™
1021003
2 hexnuts and 2 ferrules
£35.20
Pack of 2
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15259103
View Documents
Trajan™
1021011
10 hexnuts and 10 ferrules
£153.00
Pack of 10
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
Description
Description
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Specifications
10-32
Stainless Steel
5000 psi