missing translation for 'onlineSavingsMsg'
Learn More

SGE™ BP20 (Wax) GC Capillary Columns

Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.

Brand:  SGE™ 054444

 View more versions of this product

Product Code. 15218683

  • £571.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications

Specifications

280°C
BP20 (WAX)
SGE™ BP20 (Wax)
Free Acids Analysis, Free Acids, Fatty Acid Methyl Esters, Aromatics Analysis, Solvents and Essential Oils
1 μm
Polar
Hydrogen Bonding or Dipole interactions
0.32 mm
G14, G15, G16, G20, G39
20°C to 260/280°C
30 m
Fused Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.