missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ GC Capillary Column Connectors: Replacement Ferrules

Replacement SilTite ferrules for SilTiteμ-Union

Brand:  Trajan™ 073571

 View more versions of this product

Product Code. 15238943

  • £132.00 / Pack of 10
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
  • Zero dead volume for optimized peak shape
  • Chemically deactivated stainless steel ferrules give system inertness
  • Robust and stable for both high temperature and pressure cycling
Specifications

Specifications

Replacement μ-Union Ferrule
Stainless Steel
0.53 mm I.D. First Column and Second Column with 0.8 mm Holes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers