Learn More
Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan™ 073572
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Replacement μ-Union | |
0.1 - 0.32 mm and 0.1 - 0.32 mm | |
GC Capillary Column Connectors |