Learn More
Trajan™ GC Capillary Column Connectors: Replacement μ-Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073574
Code : NEW
Additional Details : Weight : 0.00090kg
 View more versions of this product
Product Code. 15278943
  • £130.00 / Pack of 5
Estimated Shipment: 05-08-2024
to see stock.
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Replacement μ-Union
0.53 mm and 0.53 mm
GC Capillary Column Connectors