Learn More
SGE™ LC Pump Priming Syringe
Product Code. 11993064
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Each
Product Code. 11993064
Brand: SGE™ 008762
For loop fill injection or sample loop flush
  • Fixed Luer lock
Specifications
For Use With (Application) Used for loop fill injection or sample loop flush
Product Type Manual Syringe
Model 5MDF-LL-GT
Quantity Ea.
Volume (Metric) 5 mL
Needle Point Style LC