missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ Thermo Scientific Split Straight-Through Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™ 092150
Code : 13
Additional Details : Weight : 0.45000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
GC Inlet Liner, Split, Straight-Through | |
105 mm | |
8 mm |
5/Pk. | |
5 mm |