Learn More
SGE™ NanoVolume Syringes
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
£85.25 - £184.00
Specifications
Accuracy | ±2% |
---|---|
Material | Borosilicate Glass (Barrel) |
Product Type | NanoVolume Syringe |
Quantity | Ea. |
Product Code | Brand | Volume (Metric) | Needle Gauge | Model | Needle Point Style | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Model | Needle Point Style | Price | Quantity & Availability | ||
10711432
|
SGE™
000300 |
0.5 μL | 23 G | 0.5BNR-5 | Removable Cone |
£179.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10370901
|
SGE™
000303 |
0.5 μL | 26 G | 0.5BNR-5/0.47 | Removable Cone |
£114.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10482001
|
SGE™
000310 |
0.5 μL | 23 G | 0.5BR-7 | Removable Cone |
£114.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11982585
|
SGE™
000311 |
0.5 μL | 23 G | 0.5BR-7BV | Removable Bevel |
£114.00
Each |
Estimated Shipment: 31-12-2024
Log in to see stock. |
||
10306392
|
SGE™
000355 |
0.5 μL | 23 G | 0.5BR-7-RAX | Removable Cone |
£153.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10095500
|
SGE™
000350 |
0.5 μL | 23 G | 0.5BNR-5-RAX | Removable Cone |
£153.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10725046
|
SGE™
000501 |
1 μL | 23 G | 1BR-5BV | Removable Bevel |
£86.85
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10636231
|
SGE™
000376 |
0.5 μL | 26 G | 0.5BR-OC-7/0.47 | Removable Bevel |
£116.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10616421
|
SGE™
000505 |
1 μL | 23 G | 1BR-7 | Removable Cone |
£182.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11992585
|
SGE™
000506 |
1 μL | 23 G | 1BR-7BV | Removable Bevel |
£114.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10167260
|
SGE™
000510 |
1 μL | 23 G | 1BR-11.5 | Removable Cone |
£89.20
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10568882
|
SGE™
000570 |
1 μL | 26 G | 1BR-7/0.47 | Removable Cone |
£85.25
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10601813
|
SGE™
000550 |
1 μL | 23 G | 1BR-5-RAX | Removable Cone |
£131.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10318983
|
SGE™
000801 |
5 μL | 23 G | 5BR-5BV | Removable Bevel |
£96.50
Each |
Estimated Shipment: 31-12-2024
Log in to see stock. |
||
10588882
|
SGE™
000802 |
5 μL | 23 G | 5BR-7 | Removable Cone |
£99.05
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11902595
|
SGE™
000803 |
5 μL | 23 G | 5BR-7BV | Removable Bevel |
£96.50
Each |
Estimated Shipment: 31-12-2024
Log in to see stock. |
||
12907536
|
SGE™
000804 |
5 μL | 23 G | 5BR-11.5 | Cone |
£96.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10578302
|
SGE™
000852 |
5 μL | 23 G | 5BR-7-RAX | Cone |
£129.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11962585
|
SGE™
000301 |
0.5 μL | 23 G | 0.5BNR-5BV | Removable Bevel |
£114.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10763714
|
SGE™
000500 |
1 μL | 23 G | 1BR-5 | Removable Cone |
£184.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10288860
|
SGE™
000800 |
5 μL | 23 G | 5BR-5 | Removable Cone |
£96.50
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
- Ability to inject down to 50nL with high precision and accuracy
- Ideal for dispensing very small liquid volumes
- Plunger extends into the needle tip
- Sample is contained only in the needle without any glass contact
- Virtually zero dead volume and carry over between injections
- Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
±2% | |
Borosilicate Glass (Barrel) | |
NanoVolume Syringe | |
Ea. |