Learn More
Trajan™ PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
Brand:  Trajan™ 0624352
 View more versions of this product
Product Code. 15218783
  • £77.60 / Pack of 5
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Robust - PEEK polymer exterior and fused silica inner tubing
  • Smooth wall surface giving excellent flow characteristics
  • Compatible with most organic solvents
  • Inertness - extremely low absorption characteristics
  • Precise ID
Specifications
Specifications
Tubing
1.5 mm
0.175 mm
100 mm