Learn More
Trajan™ PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
Brand: Trajan™ 0624353
This item is not returnable.
View return policy
Description
- Robust - PEEK polymer exterior and fused silica inner tubing
- Smooth wall surface giving excellent flow characteristics
- Compatible with most organic solvents
- Inertness - extremely low absorption characteristics
- Precise ID
Specifications
Tubing | |
0.0625 in. | |
200 mm |
1.5 mm | |
0.175 mm |