missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ PerkinElmer AutoSystem and Clarus 500 Liners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan™ 092103

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10527152

  • £126.00 / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with PerkinElmer instruments
Specifications

Specifications

AutoSystem and Clarus 500 liner, Splitless Straight Through
92 mm
6.2 mm
5/Pk.
2 mm
Autosystem Clarus 500
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers