Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Brand:  Trajan™ 039871
 View more versions of this product
Product Code. 11909437
  • £58.55 / Pack of 5
Explore more special offers
Description
Description
Designed for optimum septum penetration and prevention of septum coring.
  • Needle length is the exposed length of needle
  • Bevel tip style
  • Supplied in packs of 5
Specifications
Specifications
16 G
1.57 mm
Needle
1.19 mm
4.52 in.