Learn More
Trajan™ Replacement Kit for Plunger-in-Needle Syringes
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
Product Code | Brand | Diameter (Metric) Outer Needle | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer Needle | For Use With (Equipment) | Price | Quantity & Availability | ||||
10137260
|
Trajan™
033010 |
0.63 mm | 6.5 mm OD Syringe (0.5μL) |
N/A
|
N/A | ||||
10107360
|
Trajan™
033012 |
0.47 mm | 6.5 mm OD Syringe (0.5μL) |
N/A
|
N/A | ||||
15248583
|
Trajan™
034056 |
0.63 mm | 8 mm OD Syringe (1μL) |
N/A
|
N/A | ||||
10686221
|
Trajan™
034610 |
0.47 mm | 8 mm OD Syringe (1μL) |
N/A
|
N/A | ||||
10656601
|
Trajan™
034059 |
0.63 mm | Syringes (1μL) |
N/A
|
N/A | ||||
Description
- High precision and accuracy for small volume injections
- Replacement plunger/needle kits
- Kits are matched. Needles and plungers must be replaced as a set