Learn More
Trajan™ Replacement Needle for 10μL Syringe
A range of quality needles to fit a SGE syringe in a range of lengths and tip styles
£38.05 - £72.20
Specifications
Product Type Replacement Needle
For Use With (Equipment) Syringes (10μL)
View More Specs
Products 3
Product Code Brand Needle Point Style Diameter (Metric) Inner Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
Product Code Brand Needle Point Style Diameter (Metric) Inner Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
15228593
View Documents
Trajan™
037011
Cone 0.11 mm 0.63 mm 50 mm
£38.05
Pack of 2
10166970
View Documents
Trajan™
037610
On-column 0.1 mm 0.17 mm 100 mm
£72.20
Each
10176970
View Documents
Trajan™
037675
On-column 0.1 mm 0.23 mm 75 mm
£68.95
Pack of 2
Description
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
Replacement Needle
Syringes (10μL)