Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Brand:  Trajan™ 0318121
 View more versions of this product
Product Code. 15278533
  • £51.85 / Pack of 2
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
PTFE (Tip)
IN VITRO use only. For lab analyzers only. Not for human use.
Replacement Plunger, 10R-C/T-GT
10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler