missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Split/Splitless Liner with Single Taper

For various GC Instruments

Brand:  SGE™ 092229

Product Code. 12964071

  • £524.00 / Pack of 25
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • All liners are deactivated using high temperature (420°C) gas phase deactivation
  • Supplied in blister packs to prevent contamination
  • Reduced active sites mean no unwanted reaction in samples

Specifications

Specifications

Single taper
7.85 cm
0.4 cm
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
25/Pk.
GC Split/Splitless Liner
0.63 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers