Learn More
Trajan™ Thermo Scientific Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073452
 View more versions of this product
Product Code. 12927101
  • £94.30 / 1 set
Estimated Shipment: 08-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
GC Interface Connection
SilTite Metal
10 ferrules, 2 nuts
0.45 to 0.53 mm