missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530349
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a role in JNK activation.Specifications
SH2D3A | |
Polyclonal | |
Unconjugated | |
SH2D3A | |
Novel SH2-containing protein 1; NSP1; SH2 domain containing 3A; SH2 domain-containing 3A; SH2 domain-containing protein 3A; SH2D3A; UNQ175/PRO201 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10045 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BRG2 | |
SH2D3A | |
Recombinant fragment corresponding to a region within amino acids 92 and 392 of Human SH2D3A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |