missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12435-1-AP-20UL
This item is not returnable.
View return policy
Specifications
SH3GL2 | |
Polyclonal | |
Unconjugated | |
SH3GL2 | |
CNSA2, EEN B1, Endophilin 1, Endophilin A1, SH3 domain GRB2 like 2, SH3 domain protein 2A, SH3D2A, SH3GL2, SH3P4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116743, 20404, 6456 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35179, Q62420, Q99962 | |
SH3GL2 | |
SH3GL2 Fusion Protein Ag3114 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SH3GL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction