Learn More
Manutan Shackles, 0.75T large D, high tensile steel
Brand:  Manutan A339998
Encompass_Preferred
Product Code. 12811849
  • £24.81 / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • High tensile steel
  • Dimensions : d - 13mm ; D - 16mm ; w - 28mm ; s - 54mm ; e - 31mm