missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Shackles, 1.5T large D, high tensile steel
Brand: Manutan A339999
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Use to securely connect lifting device to the load to be lifted.- Large Dee - Bolt
- High tensile steel
- Dimensions : d - 16mm ; D - 19mm ; w - 31mm ; s - 64mm ; e - 38mm