Learn More
Manutan Shackles, 3.25T SWL D, galvanized alloy steel
Brand:  Manutan A340004
Encompass_Preferred
Product Code. 12858768
  • £18.45 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • Galvanised alloy steel
  • Dimensions : d - 16mm ; D - 19mm ; w - 26mm ; s - 51mm ; e - 38mm