Learn More
Manutan Shackles, 3.75T large bow, high tensile steel
Brand:  Manutan A340015
Encompass_Preferred
Product Code. 12812768
  • £43.26 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • High tensile steel
  • Dimensions : d - 25mm ; D - 28mm ; w - 44mm ; 2r - 63mm ; s - 107mm ; e - 56mm