Learn More
Grant Instruments™ Compact Incubator Shaker
ES-20 Versatile and programmable bench-top orbital shaker-incubator for mixing and incubating biological cultures and samples in a variety of flasks and vessels.
Brand: Grant Instruments™ ES-20
1792.22 GBP valid until 2024-12-31
Use promo code "21970" to get your promotional price.
Code : 04
Additional Details : Weight : 14.50000kg
Includes: Incubator Shaker only. (Accessories such as platforms, plastic bath tanks, flasks, and vessels are sold separately.)
Description
- Versatile programmable bench-top shaker-incubator for mixing and incubating biological fluids, samples, cell cultures and tissues
- Digital control of time, temperature and shaking speed for accuracy and repeatability
- Robust, compact construction with clear 7mm thick Plexiglass panels for easy visibility of chamber contents
- Designed for easy assembly/ disassembly and easy to move from one location to another
- Ideal for growing cell cultures in flasks and other vessels and for other general sample preparation applications
Specifications
10 mm | |
50 to 250 rpm | |
120/230 V | |
340 x 340 x 435 mm | |
340 mm | |
1 min. to 96 hr. |
2.5 kg | |
230 V, 50/60 Hz | |
25°C to 42°C | |
435mm | |
15 kg |