missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Adson Fine Serrated Thumb Forceps, Straight, standard, 4.75 in. (12.1cm)

Epredia™ Shandon Adson Fine Serrated Thumb Forceps are available in premium and standard grades.

£59.38

Specifications

Shape Straight
Length (English) 4.75 in.
Length (Metric) 12.1 cm
View More Specs

Products 1
Product Code Brand Grade Price Quantity & Availability  
Product Code Brand Grade Price Quantity & Availability  
15604454
View Documents
Epredia™
7626
Premium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Features:
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
Specifications

Specifications

Straight
4.75 in.
12.1 cm
Videos
SDS
Documents

Documents

Product Certifications