Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
Brand:  Epredia™ B1000730BL
Code : 04
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 12593926
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Writing face is flat 45° angle
  • Locking lid attached by two hinges
  • Front or back opening
Ordering Information:
  • Packaged in 75-cassette tubes
Specifications
Specifications
Small Square Pore
45°
Blue
Biopsy Cassette