Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Aid the process of speedy and reliable biopsy specimen labeling with Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
Brand: Epredia™ B1000730TN
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Writing face is flat 45° angle
- Locking lid attached by two hinges
- Front or back opening
Ordering Information:
- Packaged in 75-cassette tubes
Specifications
Small Square Pore | |
45° | |
Tan | |
Biopsy Cassette |