Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II, White
Hold biopsy specimens during processing and embedding with the special design of Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II.
£89.76 - £106.63
Specifications
Pore Style Small Square Pore
Writing Face Angle 45°
Material Acetal Polymer
Product Type Biopsy Cassette
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12700928
View Documents
Epredia™
1001097
White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12710928
View Documents
Epredia™
1001105
Lilac This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Acetal polymer construction
  • Locking lid attached by two hinges
  • Cassette writing face is at a 45° angle
  • Pore style is small square
  • Variety of colors available
Ordering Alerts:
  • Supplied in boxes of 500 cassettes with attached lids
Specifications
Specifications
Small Square Pore
45°
Acetal Polymer
Biopsy Cassette