Learn More
Shandon™ Consul No-Stick Coverslips
Optimize automated coverslipping with these coverslips, designed to not stick together and ideal for use on the Thermo Scientific Shandon Consul Automated Coverslipper.
Brand:  Epredia 7411282
Product Code. 12608016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Optimize automated coverslipping with Thermo Scientific™ Shandon™ Consul No-Stick Coverslips, designed to not stick together and ideal for use on the Thermo Scientific Shandon Consul Automated Coverslipper.
Ordering Informtion
10 x 100 pieces per pack
Specifications
Specifications
24 mm
#1.5 (0.16 to 0.19 mm)
L x W x D: 24 x 55 x 1.5mm
55 mm
55 x 24 mm
Thermo Scientific Shandon Consul Automated Coverslipper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Shandon™ Consul No-Stick Coverslips >