missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ CytoRich™ Red Collection Fluid
Allow for more accurate evaluation of cytological features with Epredia™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
£199.68 - £596.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12628346
|
Epredia™
B9990802 |
20 mL |
£596.00
Pack of 180 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12638346
|
Epredia™
B9990803 |
120 mL |
£329.00
Pack of 125 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12502716
|
Epredia™
B9990801 |
500 mL |
£199.68
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15337908
|
Epredia™
B9990800 |
1 gal. |
£236.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Pre-filled containers available
- Ready-to-use, alcohol-based fluid
- Compatible with immunohistochemistry
Specifications
Collection Fluid |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 2
- Flammable liquid Category 3
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H361-Suspected of damaging fertility or the unborn child.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol, methyl alcohol, ethylene glycol, formaldehyde, sodium hydroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Shandon™ CytoRich™ Red Collection Fluid