Learn More
Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
Brand:  Epredia™ 9990323
 View more versions of this product
Product Code. 12570057
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Specifications
Collection Fluid
120mL
Alcohol/Carbowax (PEG) combination